Chloropentacarbonylrhenium(I), 98%
Chloropentacarbonylrhenium(I), 98%
Chloropentacarbonylrhenium(I), 98%
Thermo Scientific Chemicals

Chloropentacarbonylrhenium(I), 98%

CAS: 14099-01-5 | C5ClO5Re | 361.707 g/mol
Have Questions?
Catalog NumberQuantity
042096.03
also known as 042096-03
1 g
Catalog number 042096.03
also known as 042096-03
Price (EUR)
173,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
173,00
Each
Specifications
CAS14099-01-5
Chemical Name or MaterialChloropentacarbonylrhenium(I)
ColorBrown to White
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECS Number237-948-1
View more
Pentacarbonylchlororhenium(I) was used to study π-electron deficient complexes between tricarbonylhalorhenium and diimine ligands a well as to prepare [3]-ferrocenophane complexes. It has also been employed in preparation of a series of mixed-valence compounds of rhenium and iron. These compounds find application in the study of charge-transfer processes. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only