Indium(III) isopropoxide, 99.9% (metals basis)
Indium(III) isopropoxide, 99.9% (metals basis)
Indium(III) isopropoxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Indium(III) isopropoxide, 99.9% (metals basis)

Precursor used in the preparation of indium oxide containing films | CAS: 38218-24-5 | C9H21InO3 | 292.082 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
Catalog number 042098.02
also known as 042098-02
Price (EUR)
57,90
Each
Quantity:
0.5 g
Request bulk or custom format
Price (EUR)
57,90
Each
Chemical Identifiers
CAS38218-24-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Total Metal Impurities≤0.1% (≤1000ppm)
Indium(III) isopropoxide acts as a precursor for the preparation of indium oxide containing films. It finds application in nano-materials and thin films display. Furthermore, it is used in imaging catalysis and in preparation of sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only