Molybdenum plate, 4.76mm (0.188in) thick, 99.95% (metals basis)
Molybdenum plate, 4.76mm (0.188in) thick, 99.95% (metals basis)
Molybdenum plate, 4.76mm (0.188in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Molybdenum plate, 4.76mm (0.188in) thick, 99.95% (metals basis)

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Catalog NumberQuantity
042143.GH100 x 100 mm
Catalog number 042143.GH
Price (EUR)
-
Quantity:
100 x 100 mm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7439-98-7
ChEBICHEBI:28685
Chemical Name or MaterialMolybdenum plate
EINECS Number231-107-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only