Sulfur in Light Mineral Oil standard solution, Specpure™, 100μg/g (0.0100%)
Thermo Scientific Chemicals

Sulfur in Light Mineral Oil standard solution, Specpure™, 100μg/g (0.0100%)

 
Have Questions?
Catalog NumberQuantity
042159.AE
also known as 042159-AE
100 mL
Catalog number 042159.AE
also known as 042159-AE
Price (EUR)
96,10
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
96,10
Each
Specifications
Chemical Name or MaterialSulfur in Light Mineral Oil standard solution
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H350
May cause cancer.
Health Hazard 3GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL NumberMFCD00804270
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only