Palladium Silver foil, 0.025mm (0.001in) thick, 99.9% (metals basis excluding Pt)
Thermo Scientific Chemicals

Palladium Silver foil, 0.025mm (0.001in) thick, 99.9% (metals basis excluding Pt)

Pd:Ag; 75:25 wt%
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
Catalog number 042187.FI
also known as 042187-FI
Price (EUR)
-
Quantity:
50 x 50 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialPalladium Silver foil
MDL NumberMFCD02091738
Molecular FormulaPd:Ag; 75:25 wt%
PackagingTube
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only