Tungsten, plasma standard solution, Specpure™ W 1000μg/mL
Tungsten, plasma standard solution, Specpure™ W 1000μg/mL
Tungsten, plasma standard solution, Specpure™ W 1000μg/mL
Thermo Scientific Chemicals

Tungsten, plasma standard solution, Specpure™ W 1000μg/mL

(NH4)2WO4 in H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 042248.AE
also known as 042248-AE
Price (EUR)
111,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
111,00
Each
Specifications
Chemical Name or MaterialTungsten, plasma standard solution, Specpure™
MDL NumberMFCD00011461
Molecular Formula(NH4)2WO4 in H2O
Physical FormLiquid
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only