Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
Thermo Scientific Chemicals

Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 042258.30
also known as 042258-30
Price (EUR)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.99% (metals basis)
Appearance (Color)Silvery white
FormSlug, 3.175mm (0.125in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only