1,1,1,3,3,3-Hexafluoro-2-propanol-d{2}, 98%(Isotopic)
1,1,1,3,3,3-Hexafluoro-2-propanol-d{2}, 98%(Isotopic)
1,1,1,3,3,3-Hexafluoro-2-propanol-d{2}, 98%(Isotopic)
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol-d{2}, 98%(Isotopic)

CAS: 38701-74-5 | C3H2F6O | 170.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 x 1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042291.46
also known as 042291-46
Price (EUR)
-
Quantity:
5 x 1 g
Request bulk or custom format
Chemical Identifiers
CAS38701-74-5
IUPAC Namehexafluoro(²H)propan-2-(²H)ol
Molecular FormulaC3H2F6O
InChI KeyBYEAHWXPCBROCE-AWPANEGFSA-N
SMILES[2H]OC([2H])(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
GC/MS Analysis≥98.0%
Purity1H NMR For Chemical Purity: ≥98.0%
Water Content≤200 ppm
Purity1H NMR for Isotopic Enrichment: ≥98.0%
1,1,1,3,3,3-Hexafluoro-2-propanol-d{2} is a stable isotope used for biochemical research. It is also used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only