Copper slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.996% (metals basis), Thermo Scientific Chemicals
Copper slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.996% (metals basis), Thermo Scientific Chemicals
Copper slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.996% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.996% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
042350.22
also known as 042350-22
100 g
Catalog number 042350.22
also known as 042350-22
Price (EUR)
61,90
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
61,90
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.996% (metals basis)
Appearance (Color)Brown
FormSlug, 6.35mm (0.25in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only