Molybdenum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis)
Molybdenum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis)
Molybdenum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis)
Thermo Scientific Chemicals

Molybdenum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis)

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 042361.09
also known as 042361-09
Price (EUR)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSlug, 3.175mm (0.125in) diameter.
Appearance (Color)Gray
Assay from Supplier's CofA≥99.95% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only