2,2,2-Trifluoroethanol-d{2}, 98% (Isotopic), Thermo Scientific Chemicals
2,2,2-Trifluoroethanol-d{2}, 98% (Isotopic), Thermo Scientific Chemicals
2,2,2-Trifluoroethanol-d{2}, 98% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trifluoroethanol-d{2}, 98% (Isotopic), Thermo Scientific Chemicals

NMR Solvent
Have Questions?
Catalog NumberQuantity
042362.03
also known as 042362-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042362.03
also known as 042362-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS132248-58-9
IUPAC Name2,2,2-trifluoroethan-1-ol
Molecular FormulaC2H3F3O
InChI KeyRHQDFWAXVIIEBN-UHFFFAOYSA-N
SMILESOCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay from Supplier's CofA≥98.0% ¹H nmr
Water Content (Karl Fischer Titration)≤ 500ppm
Assay (GC)≥98.0%
FormLiquid
View more
NMR Solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only