Thermo Scientific Chemicals

Poly(2-ethyl-2-oxazoline), M.W. 50,000, Thermo Scientific Chemicals

Catalog number: 042396.36
500 g, Each
Thermo Scientific Chemicals

Poly(2-ethyl-2-oxazoline), M.W. 50,000, Thermo Scientific Chemicals

Catalog number: 042396.36
500 g, Each
Quantity
Catalog number: 042396.36
also known as 042396-36
Price (EUR)
Price: 136,00
Online price: 122,40
Your price:
Quantity
-

Chemical Identifiers

CAS
25805-17-8
Molecular Formula
(C5H9NO)n
SMILES
CCC(=O)N(-*)CC-*
MDL Number
MFCD00134377
Loss on Drying
2%, max ( 1g sample, 2hours @ 125 deg C)
Appearance (Color)
Off-white to pale yellow
Form
Granular solid
Viscosity
5 to 8 cSt ( Kinematic viscosity, 10% aq. solution at 100 degree F)

Description

Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text