N,N-Dimethylacrylamide, 98%, stab. with 100ppm 4-methoxyph enol
N,N-Dimethylacrylamide, 98%, stab. with 100ppm 4-methoxyph enol
N,N-Dimethylacrylamide, 98%, stab. with 100ppm 4-methoxyph enol
Thermo Scientific Chemicals

N,N-Dimethylacrylamide, 98%, stab. with 100ppm 4-methoxyph enol

CAS: 2680-03-7 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 042405.18
also known as 042405-18
Price (EUR)
50,30
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
50,30
Each
Chemical Identifiers
CAS2680-03-7
IUPAC NameN,N-dimethylprop-2-enamide
Molecular FormulaC5H9NO
InChI KeyYLGYACDQVQQZSW-UHFFFAOYSA-N
SMILESCN(C)C(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Suppliers CofA≥97.5%
Refractive Index1.4650-1.4750
Stabilizer50-150 ppm MEHQ
Appearance (Color)Clear colorless
N,N-Dimethylacrylamide is a useful monomer for synthetic fibers and plastics. It is also used in the synthesis of photoresponsive polymers and copolymers by grafting polysiloxane on the side chain of DMA by homopolymerization or grafting cellulose to form cellulose polymers and copolymerization of methyl methacrylate with DMA or copolymerization of DMA with styrene and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only