Buffer solution, pH 1.68 (+/-0.026 @ 25°C), No Color, Specpure, NIST Traceable
Buffer solution, pH 1.68 (+/-0.026 @ 25°C), No Color, Specpure, NIST Traceable
Buffer solution, pH 1.68 (+/-0.026 @ 25°C), No Color, Specpure, NIST Traceable
Thermo Scientific Chemicals

Buffer solution, pH 1.68 (+/-0.026 @ 25°C), No Color, Specpure, NIST Traceable

CAS: 7447-40-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 042415.AP
also known as 042415-AP
Price (EUR)
32,00
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
32,00
Each
Specifications
CAS7447-40-7
CAS7647-01-0
CAS7732-18-5
Chemical Name or MaterialBuffer solution
ColorColorless
View more
Buffer Solution pH 1.68 used as a means of keeping pH at a nearly constant value in a different chemical applications. Glycine buffer solution pH 2 used for eluting antibodies in various antibody purification procedures i.e. protein A-agarose, protein G-agarose, affinity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only