Tin, Oil based standard solution, Specpure™ Sn 5000μg/g
Tin, Oil based standard solution, Specpure™ Sn 5000μg/g
Tin, Oil based standard solution, Specpure™ Sn 5000μg/g
Thermo Scientific Chemicals

Tin, Oil based standard solution, Specpure™ Sn 5000μg/g

 
Have Questions?
Catalog NumberQuantity
042450.18
also known as 042450-18
50 g
Catalog number 042450.18
also known as 042450-18
Price (EUR)
106,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
106,00
Each
Specifications
Chemical Name or MaterialTin, Oil based standard solution
Health Hazard 1H304
Health Hazard 3P301+P310-P331-P501c
MDL NumberMFCD00133862
Name NoteSn 5000μg/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only