Lanthanum(III) 2-methoxyethoxide, 99.9% (REO), 5% w/v in 2-methoxyethanol
Thermo Scientific Chemicals

Lanthanum(III) 2-methoxyethoxide, 99.9% (REO), 5% w/v in 2-methoxyethanol

Preparation of lanthanum oxide thin films by the Sol-Gel method | CAS: 115685-56-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 042522.AE
also known as 042522-AE
Price (EUR)
652,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
652,00
Each
Specifications
Assay Percent Notes(REO)
CAS115685-56-8
Chemical Name or MaterialLanthanum (III) 2-methoxyethoxide
ColorYellow
Concentration or Composition (by Analyte or Components)5% w/v in 2-methoxyethanol
View more
Lanthanum(III) 2-methoxyethoxide is used in the preparation of lanthanum oxide thin films by the Sol-Gel method

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only