Calcium hypochlorite, tech.
Calcium hypochlorite, tech.
Calcium hypochlorite, tech.
Thermo Scientific Chemicals

Calcium hypochlorite, tech.

For Use in Deodorant, Oxidizing Agent, Bleaching Agent | CAS: 7778-54-3 | CaCl2O2 | 142.976 g/mol
Have Questions?
Catalog NumberQuantity
042548.30
also known as 042548-30
250 g
Catalog number 042548.30
also known as 042548-30
Price (EUR)
49,90
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
49,90
Each
Chemical Identifiers
CAS7778-54-3
IUPAC Namecalcium dihypochlorite
Molecular FormulaCaCl2O2
InChI KeyZKQDCIXGCQPQNV-UHFFFAOYSA-N
SMILES[Ca++].[O-]Cl.[O-]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale grey
FormGranular powder
Assay (Iodometric Titration)62-72% available chlorine
Deodorant, oxidizing agent, bleaching agentCalcium hypochlorite is used for swimming pool sanitation and other water treatment plants. It is also used to disinfect drinking water, equipments in the industry and surfaces and equipments in kitchens. Further, it is used to bleach textiles and paper. It is also employed in bathroom cleaners and laundry detergents. It is used as an oxidizing agent in organic chemistry. It is involved in the cleavage of glycols, alfa-hydroxy carboxylic acids and keto acids to yield fragmented aldehydes or carboxylic acids respectively. It plays an important role in the haloform reaction to prepare chloroform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only