Platinum Iridium wire, 1.0mm (0.04in) dia
Thermo Scientific Chemicals

Platinum Iridium wire, 1.0mm (0.04in) dia

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 cm
25 cm
100 cm
Catalog number 042678.BU
also known as 042678-BU
Price (EUR)
-
Quantity:
25 cm
Request bulk or custom format
Specifications
Avg. Mol. Wt. or Mol. Wt. RangePt:Ir; 90:10 wt%
Chemical Name or MaterialPlatinum Iridium wire
MDL NumberMFCD02091683
Name Note1.0mm (0.04 in.) dia.
OdorOdorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only