Potassium chloride, 0.117M, Conductivity Standard
Potassium chloride, 0.117M, Conductivity Standard
Potassium chloride, 0.117M, Conductivity Standard
Thermo Scientific Chemicals

Potassium chloride, 0.117M, Conductivity Standard

CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number 042696.AP
also known as 042696-AP
Price (EUR)
39,60
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
39,60
Each
Specifications
Assay Percent Range0.117M
CAS7447-40-7
CAS7732-18-5
ChEBICHEBI:32588
Chemical Name or MaterialPotassium chloride
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only