Triethylmethylammonium chloride, 98%
Triethylmethylammonium chloride, 98%
Triethylmethylammonium chloride, 98%
Thermo Scientific Chemicals

Triethylmethylammonium chloride, 98%

CAS: 10052-47-8 | C7H18ClN | 151.678 g/mol
Have Questions?
Catalog NumberQuantity
042709.18
also known as 042709-18
50 g
Catalog number 042709.18
also known as 042709-18
Price (EUR)
81,80
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
81,80
Each
Chemical Identifiers
CAS10052-47-8
IUPAC Nametriethyl(methyl)azanium chloride
Molecular FormulaC7H18ClN
InChI KeyNIUZJTWSUGSWJI-UHFFFAOYSA-M
SMILES[Cl-].CC[N+](C)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Suppliers CofA≥97.5%
FormCrystalline powder
Effective phase transfer catalyst

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only