Polystyrene latex microsphere, 20.0 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 20.0 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 20.0 micron, 2.5 wt% dispersion in water
Thermo Scientific Chemicals

Polystyrene latex microsphere, 20.0 micron, 2.5 wt% dispersion in water

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
042719.AA
also known as 042719-AA
5 mL
Catalog number 042719.AA
also known as 042719-AA
Price (EUR)
599,00
Each
Quantity:
5 mL
Request bulk or custom format
Price (EUR)
599,00
Each
Specifications
CAS9003-53-6
ChEBICHEBI:27452
Chemical Name or MaterialPolystyrene latex microsphere
ColorWhite
Concentration or Composition (by Analyte or Components)2.5 wt% dispersion in water
View more
Polystyrene latex microsphere is used as a biocompatible material, especially in dentistry. They are used as insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads. It is also used as extrusion molded foamed packaging, dinnerware, bottles, toys and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only