Potassium hydroxide, 0.5N Standardized Solution in methanol
Potassium hydroxide, 0.5N Standardized Solution in methanol
Potassium hydroxide, 0.5N Standardized Solution in methanol
Thermo Scientific Chemicals

Potassium hydroxide, 0.5N Standardized Solution in methanol

CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantity
042735.K2
also known as 042735-K2
1 L
Catalog number 042735.K2
also known as 042735-K2
Price (EUR)
48,90
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
48,90
Each
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
FormLiquid
Assay from Suppliers CofA0.500N ± 0.14%
Potassium hydroxide, 0.5N standardized solution in methanol is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only