Lithium iodide, ultra dry, 99.999% (metals basis)
Lithium iodide, ultra dry, 99.999% (metals basis)
Lithium iodide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Lithium iodide, ultra dry, 99.999% (metals basis)

Light Sensitive and Hygroscopic | CAS: 10377-51-2 | ILi | 133.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 042851.09
also known as 042851-09
Price (EUR)
227,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
227,00
Each
Chemical Identifiers
CAS10377-51-2
IUPAC Namelithium(1+) iodide
Molecular FormulaILi
InChI KeyHSZCZNFXUDYRKD-UHFFFAOYSA-M
SMILES[Li+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities0.001% max.
Appearance (Color)White
Lithium iodide is used as a absorbent in refrigeration process, as a catalyst, and in photography. It is also useful for organic synthesis, pessat iodination, cleavage of esters, ethers, epoxides, decarboethoxylation, carbonylation, and aldol condensation. It is used for manufacturing medicines and mineral waters. It is used in electochemicstry and redox reactions. Lithium iodide is also a catalyzed conjugate addition reaction of β-dicarbonyl compounds. It is also useful for tetrathioquinodimethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only