Platinum 13 wt% Rhodium wire, 0.076mm (0.003in) dia, ISA Type R Standard Grade Thermocouple
Thermo Scientific Chemicals

Platinum 13 wt% Rhodium wire, 0.076mm (0.003in) dia, ISA Type R Standard Grade Thermocouple

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 cm
2 m
Catalog number 042902.BW
also known as 042902-BW
Price (EUR)
-
Quantity:
50 cm
Request bulk or custom format
Specifications
Avg. Mol. Wt. or Mol. Wt. RangePt:Rh; 87:13 wt%
Chemical Name or MaterialPlatinum 13 wt% Rhodium wire
MDL NumberMFCD00198186
Name Note0.076mm (0.003 in.) dia., ISA Type R Standard Grade Thermocouple
OdorOdorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only