Tetraethylammonium hydroxide, 35% w/w aq. soln., Reagent Grade
Tetraethylammonium hydroxide, 35% w/w aq. soln., Reagent Grade
Tetraethylammonium hydroxide, 35% w/w aq. soln., Reagent Grade
Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 35% w/w aq. soln., Reagent Grade

CAS: 77-98-5 | C8H21NO | 147.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 043023.22
also known as 043023-22
Price (EUR)
70,70
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
70,70
Each
Chemical Identifiers
CAS77-98-5
IUPAC Nametetraethylazanium hydroxide
Molecular FormulaC8H21NO
InChI KeyLRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES[OH-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormClear liquid
Assay (Titration)34.5 to 35.5%
Residual halide1000 ppm, max ( as Chloride)
Trace MetalK+Na: 20 ppm, max
Color (APHA)60, max
View more
Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only