Germanium(IV) ethoxide, 99.995% (metals basis)
Germanium(IV) ethoxide, 99.995% (metals basis)
Germanium(IV) ethoxide, 99.995% (metals basis)
Thermo Scientific Chemicals

Germanium(IV) ethoxide, 99.995% (metals basis)

Preparation of GeO2 by CVD; intermediate in sol-gel synthesis | CAS: 14165-55-0 | C8H20GeO4
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 043052.06
also known as 043052-06
Price (EUR)
181,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
181,00
Each
Specifications
Assay Percent Notes(metals basis)
CAS14165-55-0
Chemical Name or MaterialGermanium (IV) ethoxide
ColorColorless
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
View more
Preparation of Germanium oxide by CVD.Sol-gel intermediate for optical fibers and photosensitive SiO2-BeO2 glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only