Thermo Scientific Chemicals

Antimony(III) ethoxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043097.06
5 g, Each
Thermo Scientific Chemicals

Antimony(III) ethoxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 043097.06
5 g, Each
Quantity
Catalog number: 043097.06
also known as 043097-06
Price (EUR)
Quantity
-

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Vapors
    Category 4
    Chronic aquatic toxicity
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    H411 - Toxic to aquatic life with long lasting effects

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P273 - Avoid release to the environment

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P391 - Collect spillage

    P501 - Dispose of contents/ container to an approved waste disposal plant