1,4-Dioxane, HPLC Grade, 99% min, packaged under Argon in resealable ChemSeal™ bottles
1,4-Dioxane, HPLC Grade, 99% min, packaged under Argon in resealable ChemSeal™ bottles
1,4-Dioxane, HPLC Grade, 99% min, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

1,4-Dioxane, HPLC Grade, 99% min, packaged under Argon in resealable ChemSeal™ bottles

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 043167.AK
also known as 043167-AK
Price (EUR)
51,90
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
51,90
Each
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid. HPLC grade, packaged under Argon in resealable ChemSeal? bottles
Specific Absorbance (E 1%/1cm)295 nm ≤ 0.01 AU
Water Content (Karl Fischer Titration)≤0.03 %
Specific Absorbance (E 1%/1cm)270 nm ≤ 0.15 AU
Residue on Evaporation≤5 ppm
View more
1,4-Dioxane HPLC Grade is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only