Ammonium hexafluorosilicate, Puratronic™, 99.999% (metals basis)
Ammonium hexafluorosilicate, Puratronic™, 99.999% (metals basis)
Ammonium hexafluorosilicate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Ammonium hexafluorosilicate, Puratronic™, 99.999% (metals basis)

CAS: 16919-19-0 | F6H8N2Si | 178.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 043184.09
also known as 043184-09
Price (EUR)
81,30
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
81,30
Each
Chemical Identifiers
CAS16919-19-0
IUPAC Namediammonium hexafluorosilanediuide
Molecular FormulaF6H8N2Si
InChI KeyITHIMUMYFVCXSL-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].F[Si--](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Ammonium hexafluorosilicate is used as a disinfectant and analytical reagent. It finds application in etching glass, metal casting, and electroplating. It is also employed as a wood preservative agent and mothproofing agent in textiles. It plays a useful application in soldering flux. It is also used to arrest caries treatment without discoloration on demineralized primary tooth enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only