Iridium wire, 0.125mm (0.005in) dia, 99.9% (metals basis)
Iridium wire, 0.125mm (0.005in) dia, 99.9% (metals basis)
Iridium wire, 0.125mm (0.005in) dia, 99.9% (metals basis)
Thermo Scientific Chemicals

Iridium wire, 0.125mm (0.005in) dia, 99.9% (metals basis)

CAS: 7439-88-5 | Ir | 192.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
50 cm
Catalog number 043208.BS
also known as 043208-BS
Price (EUR)
-
Quantity:
10 cm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7439-88-5
ChEBICHEBI:49666
Chemical Name or MaterialIridium wire
EINECS Number231-095-9
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only