Copper Nickel wire, 0.075mm (0.003in) dia
Thermo Scientific Chemicals

Copper Nickel wire, 0.075mm (0.003in) dia

Resistance alloy, negative lead for type J thermocouple
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 m
500 m
Catalog number 043269.H4
also known as 043269-H4
Price (EUR)
38,65
Online exclusive
42,30
Save 3,65 (9%)
Each
Quantity:
100 m
Request bulk or custom format
Price (EUR)
38,65
Online exclusive
42,30
Save 3,65 (9%)
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeCu:Ni 55:45 wt%
Chemical Name or MaterialCopper Nickel wire
MDL NumberMFCD00801098
Name Note0.075mm (0.003 in.) dia.
Recommended StorageAmbient temperatures
View more
Resistance alloy, negative lead for type J thermocouple

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only