Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min
Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min
Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min
Thermo Scientific Chemicals

Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min

CAS: 10025-83-9 | Cl3Ir | 298.567 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 043380.03
also known as 043380-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS10025-83-9
IUPAC Nameiridium(3+) trichloride
Molecular FormulaCl3Ir
InChI KeyDANYXEHCMQHDNX-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ir+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Assay (unspecified)Ir 63.9% min
Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only