Copper powder, spherical, -170+400 mesh, 99.9% (metals basis), Oxygen <1000ppm
Copper powder, spherical, -170+400 mesh, 99.9% (metals basis), Oxygen <1000ppm
Copper powder, spherical, -170+400 mesh, 99.9% (metals basis), Oxygen <1000ppm
Thermo Scientific Chemicals

Copper powder, spherical, -170+400 mesh, 99.9% (metals basis), Oxygen <1000ppm

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 043385.22
also known as 043385-22
Price (EUR)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, -170 to +400 mesh
Impurity contentOxygen: ≤1000 ppm
Total Metal Impurities≤0.1%
Particle SizeReport Mesh Size Results
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only