Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%
Thermo Scientific Chemicals

Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%

CAS: 32740-79-7 | O2Ru | 133.07 g/mol
Have Questions?
Catalog NumberQuantity
043403.03
also known as 043403-03
1 g
Catalog number 043403.03
also known as 043403-03
Price (EUR)
93,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
93,30
Each
Chemical Identifiers
CAS32740-79-7
IUPAC Namedioxoruthenium
Molecular FormulaO2Ru
InChI KeyWOCIAKWEIIZHES-UHFFFAOYSA-N
SMILESO=[Ru]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max., Ru 54-60%
Chemical catalyst, important material for producing resistance and capacitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only