N,N-Dimethylformamide, anhydrous, amine free, 99.9%
N,N-Dimethylformamide, anhydrous, amine free, 99.9%
N,N-Dimethylformamide, anhydrous, amine free, 99.9%
Thermo Scientific Chemicals

N,N-Dimethylformamide, anhydrous, amine free, 99.9%

Suitable For Nucleic Acids, Peptide Synthesis | CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Catalog NumberQuantity
043465.AP500 mL
Catalog number 043465.AP
Price (EUR)
47,65
온라인 행사
52,30
Save 4,65 (9%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
47,65
온라인 행사
52,30
Save 4,65 (9%)
Each
Chemical Identifiers
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥99.9%
Impurity contentAldehyde (as HCHO): ≤0.002%
Impurity contentAmines: ≤10 ppm
View more
Suitable for nucleic acids, peptide synthesisN, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only