N,N'-Methylenebisacrylamide, 99+%
N,N'-Methylenebisacrylamide, 99+%
N,N'-Methylenebisacrylamide, 99+%
Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 99+%

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 043701.14
also known as 043701-14
Price (EUR)
50,40
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
50,40
Each
Chemical Identifiers
CAS110-26-9
IUPAC NameN-[(prop-2-enamido)methyl]prop-2-enamide
Molecular FormulaC7H10N2O2
InChI KeyZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILESC=CC(=O)NCNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 99.0%
Bisacrylamide is used in biochemistry as a preparatory agent for polyacrylamide gel for electrophoretic separation of nucleic acids and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only