Polyvinylpyrrolidone, M.W. 1,300,000
Polyvinylpyrrolidone, M.W. 1,300,000
Polyvinylpyrrolidone, M.W. 1,300,000
Thermo Scientific Chemicals

Polyvinylpyrrolidone, M.W. 1,300,000

CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 043728.30
also known as 043728-30
Price (EUR)
91,60
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
91,60
Each
Chemical Identifiers
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to off-white
Assay (Titration)Vinylpyrrolidone Monomer: ≤300 ppm
Water Content≤5.0%
Assay (unspecified)K value (1% in water): 88-100
Polyvinylpyrrolidone is used to make hair sprays, shampoos, plasma substitutes, detergents and adhesives. It is also used in hair dyes, clarifying agents in wine, beer, and juices, thickeners in inks and paints, water binding agents and protective colloids. It finds application as a special additive for batteries, ceramics, fiberglass, inks, and inkjet, an inhibitor of recrystallisation and to increase resolution in photoresists for cathode ray tubes. Further, it is used for gene delivery. In addition to this, it is used as a pharmaceutical aid, complexing agent and solubilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only