Ethyl N-phenylformimidate, 97%
Ethyl N-phenylformimidate, 97%
Ethyl N-phenylformimidate, 97%
Thermo Scientific Chemicals

Ethyl N-phenylformimidate, 97%

CAS: 6780-49-0 | C9H11NO | 149.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 043788.06
also known as 043788-06
Price (EUR)
26,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
26,00
Each
Chemical Identifiers
CAS6780-49-0
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5250-1.5300 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only