Aluminum oxide-silicon oxide (13%), catalyst support, low surface area, macroporous
Aluminum oxide-silicon oxide (13%), catalyst support, low surface area, macroporous
Aluminum oxide-silicon oxide (13%), catalyst support, low surface area, macroporous
Thermo Scientific Chemicals

Aluminum oxide-silicon oxide (13%), catalyst support, low surface area, macroporous

Form: S.A. 0.008m2/g | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 043863.A1
Quantity:
1 kg
Request bulk or custom format
Specifications
AppearanceWhite spheres
CAS1344-28-1
Chemical Name or MaterialAluminum oxide
ColorWhite
IUPAC Namedialuminium(3+) trioxidandiide
View more
Aluminum oxide-silicon oxide is a fibrous material used as loose wool, blanket, felt, paper or boards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only