Platinum, 5% on activated carbon powder, sulfided, 0.5% S (as sulfide)
Platinum, 5% on activated carbon powder, sulfided, 0.5% S (as sulfide)
Platinum, 5% on activated carbon powder, sulfided, 0.5% S (as sulfide)
Thermo Scientific Chemicals

Platinum, 5% on activated carbon powder, sulfided, 0.5% S (as sulfide)

CAS: 7440-6-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 043905.09
also known as 043905-09
Price (EUR)
-
Quantity:
10 g
Request bulk or custom format
Specifications
CAS6-6-7440
Chemical Name or MaterialPlatinum
DOT InformationTransport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only