Sodium tetrachloropalladate(II) hydrate, Premion™, 99.999% (metals basis), Pd 30% min
Sodium tetrachloropalladate(II) hydrate, Premion™, 99.999% (metals basis), Pd 30% min
Sodium tetrachloropalladate(II) hydrate, Premion™, 99.999% (metals basis), Pd 30% min
Thermo Scientific Chemicals

Sodium tetrachloropalladate(II) hydrate, Premion™, 99.999% (metals basis), Pd 30% min

CAS: 13820-53-6 | Cl4Na2Pd | 294.20 g/mol
Have Questions?
Catalog NumberQuantity
043945.03
also known as 043945-03
1 g
Catalog number 043945.03
also known as 043945-03
Price (EUR)
194,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
194,00
Each
Chemical Identifiers
CAS13820-53-6
IUPAC Namedisodium tetrachloropalladiumbis(ylium)
Molecular FormulaCl4Na2Pd
InChI KeyABKQFSYGIHQQLS-UHFFFAOYSA-J
SMILES[Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max., Pd 30% min
Sodium tetrachloropalladate is used in organ metallic chemistry for the preparation of phosphine complexes of palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only