Holmium rod, 6.35mm (0.25in) dia, 99.8% (metals basis)
Holmium rod, 6.35mm (0.25in) dia, 99.8% (metals basis)
Holmium rod, 6.35mm (0.25in) dia, 99.8% (metals basis)
Thermo Scientific Chemicals

Holmium rod, 6.35mm (0.25in) dia, 99.8% (metals basis)

CAS: 7440-60-0 | Ho | 164.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
50 mm
Catalog number 044050.DJ
also known as 044050-DJ
Price (EUR)
1.104,00
Each
Quantity:
50 mm
Request bulk or custom format
Price (EUR)
1.104,00
Each
Chemical Identifiers
CAS7440-60-0
IUPAC Nameholmium
Molecular FormulaHo
InChI KeyKJZYNXUDTRRSPN-UHFFFAOYSA-N
SMILES[Ho]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged in mylar under argon
RUO – Research Use Only