Titanium Thinfoil, 0.0127mm (0.0005 in.) thick, 99.6% (metals basis), Thermo Scientific Chemicals
Titanium Thinfoil, 0.0127mm (0.0005 in.) thick, 99.6% (metals basis), Thermo Scientific Chemicals
Titanium Thinfoil, 0.0127mm (0.0005 in.) thick, 99.6% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium Thinfoil, 0.0127mm (0.0005 in.) thick, 99.6% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
044243.6N90 x 100 mm
Catalog number 044243.6N
Price (EUR)
-
Quantity:
90 x 100 mm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7440-32-6
ChEBICHEBI:33341
Chemical Name or MaterialTitanium Thinfoil
EINECS Number231-142-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only