Hafnium dinitrate oxide, 99.9% (metals basis excluding Zr), Zr <0.1%, 10% w/v aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium dinitrate oxide, 99.9% (metals basis excluding Zr), Zr <0.1%, 10% w/v aq. soln., Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
044246.AD
also known as 044246-AD
50 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044246.AD
also known as 044246-AD
Price (EUR)
-
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS36125-38-9
SpecificationsSpecification SheetSpecification Sheet
Impurity content0.1% Zr max.
Total Metal Impurities excluding Zr0.1% max.
Concentration (w/v)10% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only