Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic, 99.9999% (metals basis)
Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic, 99.9999% (metals basis)
Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic, 99.9999% (metals basis)
Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic, 99.9999% (metals basis)
Thermo Scientific Chemicals

Aluminum foil, 0.5mm (0.02in) thick, annealed, Puratronic, 99.9999% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Catalog number 044332.FI
also known as 044332-FI
Price (EUR)
103,65
Online exclusive
115,00
Save 11,35 (10%)
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (EUR)
103,65
Online exclusive
115,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only