Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III), 97%
Thermo Scientific Chemicals

Tris[N,N-bis(trimethylsilyl)amide]lanthanum(III), 97%

CAS: 35788-99-9 | C18H54LaN3Si6 | 620.066 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 044381.03
also known as 044381-03
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS35788-99-9
SpecificationsSpecification SheetSpecification Sheet
Complexometric TitrationLa: 21.7-23.0 wt%
Assay from Supplier's CofALa: ≥99.9% (metals basis)
Complexometric Titration≥96%
Appearance (Color)White to pale cream
FormCrystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only