Lithium hydroxide, anhydrous, 99.995% (metals basis)
Lithium hydroxide, anhydrous, 99.995% (metals basis)
Lithium hydroxide, anhydrous, 99.995% (metals basis)
Thermo Scientific Chemicals

Lithium hydroxide, anhydrous, 99.995% (metals basis)

CAS: 1310-65-2 | HLiO | 23.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 044471.06
also known as 044471-06
Price (EUR)
57,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
57,40
Each
Chemical Identifiers
CAS1310-65-2
IUPAC Namelithium(1+) hydroxide
Molecular FormulaHLiO
InChI KeyWMFOQBRAJBCJND-UHFFFAOYSA-M
SMILES[Li+].[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormGranules or pellets
Assay (special)?2.5 % of Theoretical value (Li)
Total Metal Impurities≤0.005% (50 ppm)
It is used as a heat transfer medium, as a storage-battery electrolyte and also used for the production of lithium greases. It is also used in ceramics, in some portland cement formulations, in the absoption of carbondioxide from sealed enviornments such as submarines, spacecrafts and breathing apparatus. It is used in esterification reactions, as stabilizer in photographic developments and as a coolant in pressurized water reactors for corrosion control.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only