Specifications
Chemical Name or MaterialTransition Metals, plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Health Hazard 2May be corrosive to metals, Causes severe skin burns and eye damage
View more