Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals
Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals
Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044558.09
Price (EUR)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS12045-63-5
IUPAC Namediboron titanium
Molecular FormulaB2Ti
InChI KeyQYEXBYZXHDUPRC-UHFFFAOYSA-N
SMILES[B].[B].[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Used as a cathode material in aluminum smelting and can be shaped by electrical discharge machining. Specialized applications in such areas as impact resistant armor, cutting tools, crucibles, neutron absorbers and wear resistant coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only