Aluminum wire, 0.076mm (0.003in) dia, hard, Puratronic™, 99.999% (metals basis)
Aluminum wire, 0.076mm (0.003in) dia, hard, Puratronic™, 99.999% (metals basis)
Aluminum wire, 0.076mm (0.003in) dia, hard, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Aluminum wire, 0.076mm (0.003in) dia, hard, Puratronic™, 99.999% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
25 m
Catalog number 044577.G5
also known as 044577-G5
Price (EUR)
104,00
Each
Quantity:
5 m
Request bulk or custom format
Price (EUR)
104,00
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver / Grey
Assay from Supplier's CofA≥99.999% (metals basis)
FormWire, 0.076mm (0.003in) diameter, hardened.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only